siRNA Details
Gene Symbol: Fosl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010235.2 | NP_034365.1 | 3 | 619 | ||
GenBank mRNA | AK144785.1 | - | 3 | 771 | ||
AK144839.1 | - | 3 | 615 | |||
AK145035.1 | - | 3 | 613 | |||
BC052917.1 | - | 3 | 581 | 30063479 | ||
U34245.1 | - | 3 | 562 |

More Information
Other Species Targeted

Gene Ontology Categories:
