siRNA Details
Gene Symbol: Fst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301373.1 | NP_001288302.1 | 4 | 1073 | ||
NM_001301375.1 | NP_001288304.1 | 4 | 1073 | |||
NM_008046.3 | NP_032072.1 | 4 | 1073 | |||
GenBank mRNA | AK079916.1 | - | 4 | 1074 | ||
AK083556.1 | - | 4 | 854 | |||
BC144926.1 | - | 4 | 897 | 9053474 | ||
BC145945.1 | - | 4 | 897 | 40131248 | ||
X83377.1 | - | 4 | 702 | |||
Z29532.1 | - | 4 | 708 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_012561, XM_006231953, XM_006231954.
Gene Ontology Categories:
