siRNA Details
Gene Symbol: Ftcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080845.2 | NP_543121.1 | 5 | 602 | ||
GenBank mRNA | AK165414.1 | - | 4 | 511 | ||
BB610765.1 | - | 603 | ||||
BC010813.1 | - | 5 | 564 | 4211065 | ||
BC024078.1 | - | 5 | 596 | 5098529 | ||
CD297973.1 | - | 5 | 605 |
