siRNA Details
Gene Symbol: HLA-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098479.1 | NP_001091949.1 | 3, 4 | 738 | ||
NM_018950.2 | NP_061823.2 | 3, 4 | 738 | |||
XM_011514564.1 | XP_011512866.1 | 3, 4 | 644 | |||
XM_017010810.1 | XP_016866299.1 | 3, 4 | 644 | |||
XM_017010811.1 | XP_016866300.1 | 3, 4 | 644 | |||
XM_017010812.1 | XP_016866301.1 | 3, 4 | 644 | |||
XM_017010813.1 | XP_016866302.1 | 3, 4 | 644 | |||
XM_017010814.1 | XP_016866303.1 | 3, 4 | 608 | |||
XM_017010815.1 | XP_016866304.1 | 2, 3 | 533 | |||
GenBank mRNA | AK096962.1 | - | 3, 4 | 644 | ||
AK125274.1 | - | 3, 4 | 390 | |||
AK225185.1 | - | 3, 4 | 642 | |||
AK298704.1 | - | 3, 4 | 644 | |||
AK300090.1 | - | 3, 4 | 677 | |||
AK301488.1 | - | 3, 4 | 625 | |||
AK302420.1 | - | 4, 5 | 608 | |||
AK303670.1 | - | 2, 3 | 700 | |||
AK309156.1 | - | 3, 4 | 644 | |||
AY216682.1 | - | 3, 4 | 602 | |||
AY221102.1 | - | 3, 4 | 602 | |||
AY253269.1 | - | 3, 4 | 614 | |||
AY253270.1 | - | 3, 4 | 614 | |||
BC009260.2 | - | 3, 4 | 670 | |||
BC062991.1 | - | 3, 4 | 618 | 5496696 | ||
DQ367723.1 | - | 3, 4 | 614 | |||
GQ891552.1 | - | 3, 4 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
