siRNA Details
Gene Symbol: Timm10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019502.2 | NP_062375.1 | 2 | 99 | ||
GenBank mRNA | AF150103.1 | - | 2 | 164 | ||
AK003368.1 | - | 2 | 73 | |||
AK003421.1 | - | 2 | 126 | |||
AK010994.1 | - | 2 | 192 | |||
BC009158.1 | - | 2 | 99 | 2648985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
