siRNA Details
Gene Symbol: HLA-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098478.1 | NP_001091948.1 | 6 | 938 | ||
NM_018950.2 | NP_061823.2 | 7 | 1214 | |||
XM_017010815.1 | XP_016866304.1 | 6 | 1009 | |||
GenBank mRNA | AK225185.1 | - | 7 | 1118 | ||
BC062991.1 | - | 7 | 1094 | 5496696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
