siRNA Details
Gene Symbol: HLA-G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002127.5 | NP_002118.1 | 4 | 620 | ||
XM_017010816.1 | XP_016866305.1 | 4 | 529 | |||
GenBank mRNA | AF071017.1 | - | 2 | 236 | ||
AF071018.1 | - | 2 | 236 | |||
AF071019.1 | - | 2 | 236 | |||
AF071020.1 | - | 2 | 236 | |||
AF196177.1 | - | 3 | 442 | |||
AF196178.1 | - | 3 | 442 | |||
AF226990.2 | - | 3 | 466 | |||
AF436089.1 | - | 2 | 369 | |||
AK093478.1 | - | 3 | 736 | |||
AK302054.1 | - | 4 | 546 | |||
AY359818.1 | - | 3 | 444 | |||
BC021708.2 | - | 4 | 529 | 4822521 | ||
DC397107.1 | - | 4 | 546 | |||
M32800.1 | - | 3 | 442 | |||
M90683.1 | - | 3 | 442 | |||
U58024.1 | - | 2 | 369 | |||
U58025.1 | - | 2 | 369 | |||
U58026.1 | - | 2 | 369 | |||
U58027.1 | - | 2 | 369 | |||
U58028.1 | - | 2 | 369 | |||
U58029.1 | - | 2 | 369 | |||
U58094.1 | - | 2 | 369 | |||
U88244.1 | - | 2 | 341 | |||
X17273.1 | - | 3 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
