siRNA Details
Gene Symbol: Fzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010246.1 | NP_034376.1 | 1 | 1018 | ||
GenBank mRNA | AF033585.2 | - | 1 | 873 | ||
AK021164.1 | - | 1 | 1202 | |||
Y17709.1 | - | 1 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
