siRNA Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010250.5 | NP_034380.1 | 3 | 935 | ||
GenBank mRNA | AK034095.1 | - | 3 | 936 | ||
AK039496.1 | - | 3 | 219 | |||
AK141596.1 | - | 3 | 795 | |||
BC132331.1 | - | 3 | 611 | 40130608 | ||
BC137733.1 | - | 3 | 614 | 8860752 | ||
M63436.1 | - | 3 | 608 | |||
M86566.1 | - | 3 | 303 | |||
X61430.1 | - | 2 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
