siRNA Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010250.5 | NP_034380.1 | 10 | 1960 | ||
GenBank mRNA | AK034095.1 | - | 10 | 1961 | ||
AK039496.1 | - | 10 | 1244 | |||
AK141596.1 | - | 10 | 1820 | |||
BC132331.1 | - | 10 | 1636 | 40130608 | ||
BC137733.1 | - | 10 | 1639 | 8860752 | ||
M63436.1 | - | 10 | 1633 | |||
M86566.1 | - | 10 | 1328 | |||
X61430.1 | - | 9 | 1216 |

More Information
Other Species Targeted

Gene Ontology Categories:
