siRNA Details
Gene Symbol: Gabra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008066.3 | NP_032092.1 | 9 | 1237 | ||
GenBank mRNA | AK039055.1 | - | 9 | 1238 | ||
AK046953.1 | - | 4 | 1371 | |||
AK048165.1 | - | 1135 | ||||
BC115727.1 | - | 9 | 1159 | 40039253 | ||
M86567.1 | - | 9 | 988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
