siRNA Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008069.4 | NP_032095.1 | 7 | 696 | ||
GenBank mRNA | BC130258.1 | - | 7 | 700 | 40142758 | |
BC157920.1 | - | 7 | 762 | 9007350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
