siRNA Details
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008076.3 | NP_032102.2 | 3, 4 | 440 | ||
GenBank mRNA | AF024621.1 | - | 3, 4 | 553 | ||
AK132642.1 | - | 3, 4 | 268 | |||
AK160446.1 | - | 1, 2 | 107 | |||
BC057957.1 | - | 3, 4 | 425 | 5357713 | ||
CK618173.1 | - | 3, 4 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
