siRNA Details
Gene Symbol: Gal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010253.3 | NP_034383.1 | 4 | 306 | ||
GenBank mRNA | AK133522.1 | - | 4 | 322 | ||
BC044055.1 | - | 4 | 306 | 6476459 | ||
Z23069.1 | - | 2 | 85 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
