siRNA Details
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008080.5 | NP_032106.1 | 8 | 1290 | ||
NM_027739.2 | NP_082015.2 | 7 | 1099 | |||
GenBank mRNA | AK040324.1 | - | 7 | 1100 | ||
AK144484.1 | - | 8 | 1256 | |||
BC019969.1 | - | 7 | 1040 | |||
BC036996.1 | - | 7 | 1024 | 5038560 | ||
BC057199.1 | - | 8 | 1221 | 5315443 | ||
L25885.1 | - | 7 | 1053 | |||
U18975.1 | - | 7 | 928 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
