siRNA Details
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010255.3 | NP_034385.1 | 3 | 444 | ||
GenBank mRNA | AF015887.1 | - | 3 | 377 | ||
AK140688.1 | - | 3 | 445 | |||
AK158087.1 | - | 3 | 387 | |||
BC049233.1 | - | 3 | 430 | 4195030 |

More Information
Other Species Targeted

Gene Ontology Categories:
