siRNA Details
Gene Symbol: Gata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008090.5 | NP_032116.4 | 6 | 1595 | ||
GenBank mRNA | AB000096.2 | - | 6 | 1487 | ||
AK004551.1 | - | 6 | 1592 | |||
AK004675.1 | - | 6 | 1924 | |||
AK154792.1 | - | 6 | 1593 | |||
AK154981.1 | - | 6 | 1596 | |||
BC107009.2 | - | 6 | 1499 | 40047573 | ||
BC107010.1 | - | 5 | 1272 | 40047574 |

More Information
Other Species Targeted

Gene Ontology Categories:
