siRNA Details
Gene Symbol: Gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008091.3 | NP_032117.1 | 3 | 1338 | ||
GenBank mRNA | AK090089.1 | - | 3 | 1347 | ||
AK155549.1 | - | 3 | 1339 | |||
AK156634.1 | - | 3 | 1325 | |||
AK157625.1 | - | 958 | ||||
AK157939.1 | - | 3 | 1212 | |||
AK161099.1 | - | 3 | 1217 | |||
BC062915.1 | - | 2 | 934 | 6826352 | ||
X55123.1 | - | 2 | 954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
