siRNA Details
Gene Symbol: Gbas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008095.4 | NP_032121.3 | 3, 4 | 330 | ||
GenBank mRNA | AJ001261.1 | - | 3, 4 | 308 | ||
AK170336.1 | - | 3, 4 | 276 | |||
BC055893.1 | - | 3, 4 | 279 | 4023066 | ||
BY292750.1 | - | 3, 4 | 331 |

More Information
Other Species Targeted

Gene Ontology Categories:
