siRNA Details
Gene Symbol: Gbp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010259.2 | NP_034389.2 | 9, 10 | 1545 | ||
GenBank mRNA | AK077796.1 | - | 9, 10 | 1547 | ||
BC108990.2 | - | 1590 | 40047769 | |||
EF494422.1 | - | 9, 10 | 1545 | |||
M55544.1 | - | 1529 | ||||
M63961.1 | - | 1623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
