siRNA Details
Gene Symbol: Gbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010260.1 | NP_034390.1 | 8 | 1366 | ||
GenBank mRNA | AF077007.1 | - | 8 | 1366 | ||
AF109168.1 | - | 8 | 1319 | |||
AJ007970.1 | - | 8 | 1359 | |||
AK150138.1 | - | 8 | 1302 | |||
AK150146.1 | - | 4 | 668 | |||
BC011336.1 | - | 8 | 1355 | 3494311 | ||
BC032882.1 | - | 8 | 1381 | 1230883 | ||
BC049141.1 | - | 8 | 1343 | |||
CT010249.1 | - | 7 | 1196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
