siRNA Details
Gene Symbol: Rabac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010261.2 | NP_034391.1 | 2 | 361 | ||
GenBank mRNA | AF120162.1 | - | 2 | 305 | ||
AF252856.1 | - | 2 | 295 | |||
AK008559.1 | - | 2 | 273 | |||
AK009726.1 | - | 2 | 221 | |||
AK046109.1 | - | 1 | 257 | |||
AK150681.1 | - | 2 | 242 | |||
AK170289.1 | - | 2 | 321 | |||
BC008242.1 | - | 2 | 294 | 2655184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
