siRNA Details
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008098.4 | NP_032124.1 | 4 | 567 | ||
GenBank mRNA | AK002620.1 | - | 4 | 481 | ||
AK028027.1 | - | 4 | 495 | |||
AK049398.1 | - | 4 | 467 | |||
AK083888.1 | - | 4 | 481 | |||
AK132029.1 | - | 4 | 490 | |||
AK145755.1 | - | 4 | 472 | |||
AK146305.1 | - | 4 | 515 | |||
AK146473.1 | - | 4 | 467 | |||
AK150750.1 | - | 4 | 489 | |||
AK151106.1 | - | 4 | 435 | |||
AK151147.1 | - | 4 | 435 | |||
AK151485.1 | - | 4 | 492 | |||
AK151564.1 | - | 4 | 435 | |||
AK151640.1 | - | 4 | 490 | |||
AK151687.1 | - | 4 | 463 | |||
AK152272.1 | - | 4 | 463 | |||
AK152317.1 | - | 4 | 452 | |||
AK194194.1 | - | 3 | 202 | |||
AK198840.1 | - | 3 | 202 | |||
AK206882.1 | - | 3 | 202 | |||
BC043084.1 | - | 4 | 464 | 6417343 | ||
BC054811.1 | - | 4 | 480 | 6839739 | ||
D78188.1 | - | 4 | 476 | |||
U20290.1 | - | 4 | 498 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_024374, U21661, BC088136, AY951952, D26179, and others.
Gene Ontology Categories:
