siRNA Details
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008098.4 | NP_032124.1 | 2, 3 | 473 | ||
GenBank mRNA | AK002620.1 | - | 2, 3 | 387 | ||
AK028027.1 | - | 2, 3 | 401 | |||
AK049398.1 | - | 2, 3 | 373 | |||
AK083888.1 | - | 2, 3 | 387 | |||
AK132029.1 | - | 2, 3 | 396 | |||
AK145755.1 | - | 2, 3 | 378 | |||
AK146305.1 | - | 2, 3 | 421 | |||
AK146473.1 | - | 2, 3 | 373 | |||
AK150750.1 | - | 2, 3 | 395 | |||
AK151106.1 | - | 2, 3 | 341 | |||
AK151147.1 | - | 2, 3 | 341 | |||
AK151485.1 | - | 2, 3 | 398 | |||
AK151564.1 | - | 2, 3 | 341 | |||
AK151640.1 | - | 2, 3 | 396 | |||
AK151687.1 | - | 2, 3 | 369 | |||
AK152272.1 | - | 2, 3 | 369 | |||
AK152317.1 | - | 2, 3 | 359 | |||
AK194194.1 | - | 1, 2 | 108 | |||
AK198840.1 | - | 1, 2 | 108 | |||
AK206882.1 | - | 1, 2 | 108 | |||
BC043084.1 | - | 2, 3 | 370 | 6417343 | ||
BC054811.1 | - | 2, 3 | 386 | 6839739 | ||
CA559569.1 | - | 2, 3 | 473 | |||
D78188.1 | - | 2, 3 | 382 | |||
U20290.1 | - | 2, 3 | 404 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_024374, U21661, BC088136, AY951952, D26179, and others.
Gene Ontology Categories:
