siRNA Details
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008101.2 | NP_032127.2 | 7 | 713 | ||
GenBank mRNA | AK004890.1 | - | 5 | 839 | ||
AK144393.1 | - | 7 | 660 | |||
BC012718.1 | - | 6 | 788 | |||
BC031885.1 | - | 7 | 663 | 5137340 | ||
BC057988.1 | - | 7 | 644 | 5100574 | ||
L38613.1 | - | 6 | 623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
