siRNA Details
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008102.3 | NP_032128.1 | 1, 2 | 451 | ||
GenBank mRNA | AK150657.1 | - | 1, 2 | 463 | ||
AK152571.1 | - | 1, 2 | 463 | |||
BC005643.1 | - | 1, 2 | 451 | 3709238 | ||
BC069921.1 | - | 1, 2 | 455 | 6529016 | ||
L09737.1 | - | 1, 2 | 438 | |||
S71373.1 | - | 1, 2 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
