siRNA Details
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010267.3 | NP_034397.1 | 2 | 233 | ||
GenBank mRNA | AK020988.1 | - | 2 | 217 | ||
AK045460.1 | - | 2 | 233 | |||
AK049655.1 | - | 2 | 203 | |||
AK083814.1 | - | 2 | 234 | |||
AK139057.1 | - | 2 | 76 | |||
AK149281.1 | - | 2 | 244 | |||
BC048177.1 | - | 2 | 209 | 6825756 | ||
BC051135.1 | - | 2 | 198 | 6532839 | ||
BC060212.1 | - | 2 | 206 | |||
Y17850.1 | - | 2 | 221 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107897.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
