siRNA Details
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010267.3 | NP_034397.1 | 6 | 925 | ||
GenBank mRNA | AK039016.1 | - | 1 | 2893 | ||
AK045460.1 | - | 6 | 925 | |||
AK083814.1 | - | 6 | 926 | |||
AK139057.1 | - | 6 | 768 | |||
AK149281.1 | - | 6 | 936 | |||
BC048177.1 | - | 6 | 901 | 6825756 | ||
BC051135.1 | - | 6 | 890 | 6532839 | ||
BC060212.1 | - | 6 | 898 | |||
Y17850.1 | - | 6 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
