siRNA Details
Gene Symbol: Gdf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010272.1 | NP_034402.1 | 2 | 690 | ||
GenBank mRNA | AF092734.1 | - | 2 | 435 | ||
KC561772.1 | - | 1802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
