siRNA Details
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002701.1 | - | 5, 6 | 908 | ||
AF079107.1 | - | 4, 5 | 921 | |||
AF079108.1 | - | 3, 4 | 837 | |||
AK133028.1 | - | 2, 3 | 183 | |||
AK138512.1 | - | 5, 6 | 1634 | |||
AK149349.1 | - | 5, 6 | 1638 | |||
AK154579.1 | - | 5, 6 | 1644 | |||
BC138561.1 | - | 5, 6 | 1029 | 8861581 | ||
CN539269.1 | - | 3, 4 | 552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
