siRNA Details
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010280.3 | NP_034410.3 | 3 | 590 | ||
GenBank mRNA | AB008833.1 | - | 3 | 468 | ||
AF020305.1 | - | 3 | 423 | |||
AF036163.1 | - | 3 | 484 | |||
AF041842.1 | - | 3 | 473 | |||
AF051766.1 | - | 3 | 485 | |||
AK046542.1 | - | 3 | 517 | |||
BC066202.1 | - | 3 | 590 | 6430904 | ||
Y15110.1 | - | 3 | 504 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053398, BC079378, AF184920.
Gene Ontology Categories:
