siRNA Details
Gene Symbol: Gip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008119.2 | NP_032145.2 | 3 | 281 | ||
GenBank mRNA | AK008308.1 | - | 3 | 281 | ||
AK008525.1 | - | 2 | 137 | |||
AK172404.1 | - | 1 | 58 | |||
BC104314.1 | - | 3 | 271 | 40044479 | ||
BC104315.1 | - | 3 | 271 | 40044480 | ||
U34295.1 | - | 2 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
