siRNA Details
Gene Symbol: Gja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008120.3 | NP_032146.1 | 2 | 903 | ||
GenBank mRNA | AF216832.1 | - | 2 | 842 | ||
AK143024.1 | - | 2 | 906 | |||
BC056613.1 | - | 2 | 837 | 4971608 |

More Information
Other Species Targeted

Gene Ontology Categories:
