siRNA Details
Gene Symbol: Gjb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010291.3 | NP_034421.1 | 2 | 616 | ||
GenBank mRNA | AK082917.1 | - | 2 | 617 | ||
BC011148.1 | - | 1 | 478 | 4207506 | ||
M91236.1 | - | 2 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
