siRNA Details
Gene Symbol: Gk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010294.1 | NP_034424.1 | 1 | 1481 | ||
GenBank mRNA | AF117734.1 | - | 1 | 1481 | ||
AK075761.1 | - | 1 | 1394 | |||
BC050764.1 | - | 1 | 1494 | 6774427 | ||
BC061147.1 | - | 1 | 1460 | 6770827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
