siRNA Details
Gene Symbol: Gclc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010295.2 | NP_034425.1 | 14 | 1839 | ||
GenBank mRNA | AK144339.1 | - | 14 | 1840 | ||
AK160876.1 | - | 14 | 1599 | |||
AK169676.1 | - | 14 | 1812 | |||
BC019374.1 | - | 14 | 1596 | 4195425 | ||
U85414.1 | - | 14 | 1564 | |||
U85498.1 | - | 14 | 1564 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_012815, BC081702, J05181.
Gene Ontology Categories:
