siRNA Details
Gene Symbol: Gclc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010295.2 | NP_034425.1 | 9 | 1251 | ||
GenBank mRNA | AK144339.1 | - | 9 | 1252 | ||
AK160876.1 | - | 9 | 1011 | |||
AK169676.1 | - | 9 | 1224 | |||
BC019374.1 | - | 9 | 1008 | 4195425 | ||
U85414.1 | - | 9 | 976 | |||
U85498.1 | - | 9 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
