siRNA Details
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008131.4 | NP_032157.2 | 4 | 552 | ||
GenBank mRNA | AK137969.1 | - | 4 | 539 | ||
AK159106.1 | - | 4 | 538 | |||
AK160670.1 | - | 4 | 539 | |||
AK162685.1 | - | 4 | 537 | |||
AK168493.1 | - | 4 | 536 | |||
AY044241.1 | - | 4 | 536 | |||
BC015086.1 | - | 4 | 536 | 4224865 | ||
U09114.1 | - | 3 | 428 | |||
X16314.1 | - | 4 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
