siRNA Details
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159626.1 | NP_001153098.1 | 2, 3 | 259 | ||
NM_024284.2 | NP_077246.2 | 2, 3 | 349 | |||
GenBank mRNA | AK147022.1 | - | 2, 3 | 310 | ||
AK150557.1 | - | 2, 3 | 267 | |||
AK151231.1 | - | 2, 3 | 267 | |||
AK152742.1 | - | 2, 3 | 267 | |||
AK165777.1 | - | 2, 3 | 253 | |||
BC004749.1 | - | 2, 3 | 292 | 3583919 | ||
BC019817.1 | - | 2, 3 | 242 | 5123648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
