siRNA Details
Gene Symbol: Glycam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289587.1 | NP_001276516.1 | 3 | 274 | ||
NM_008134.3 | NP_032160.1 | 3 | 274 | |||
GenBank mRNA | AK021358.1 | - | 3 | 272 | ||
BC031931.1 | - | 1 | 1726 | |||
BC145548.1 | - | 3 | 241 | 9054112 | ||
BC145832.1 | - | 3 | 247 | 40131135 | ||
BC145858.1 | - | 3 | 247 | 40131161 | ||
M93428.1 | - | 3 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
