siRNA Details
Gene Symbol: Gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010299.3 | NP_034429.1 | 3, 4 | 610 | ||
GenBank mRNA | AK149642.1 | - | 3, 4 | 503 | ||
AK152256.1 | - | 3, 4 | 502 | |||
AK210428.1 | - | 1, 2 | 187 | |||
BC004651.1 | - | 3, 4 | 478 | 3482848 | ||
CT010289.1 | - | 3, 4 | 431 | |||
L19526.1 | - | 3, 4 | 444 | |||
U09816.1 | - | 3, 4 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
