siRNA Details
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010308.3 | NP_034438.1 | 8 | 1443 | ||
GenBank mRNA | AK030718.1 | - | 1 | 1140 | ||
AK035907.1 | - | 8 | 1444 | |||
AK047197.1 | - | 8 | 1447 | |||
BC051989.1 | - | 8 | 1270 | 5686124 | ||
M36777.1 | - | 8 | 966 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017327, XM_008772322, XM_008772320, M12671, AF413211, and others.
Gene Ontology Categories:
