siRNA Details
Gene Symbol: Gnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008141.3 | NP_032167.1 | 6 | 648 | ||
GenBank mRNA | AK009388.1 | - | 5 | 532 | ||
AK028395.1 | - | 2 | 165 | |||
BC016272.1 | - | 6 | 646 | 4506728 | ||
BC085102.1 | - | 4 | 1395 | |||
BC131941.1 | - | 6 | 643 | 40130218 | ||
BC131943.1 | - | 6 | 643 | 40130220 | ||
L10666.1 | - | 5 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
