siRNA Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010312.4 | NP_034442.1 | 8 | 829 | ||
GenBank mRNA | AK002604.1 | - | 8 | 825 | ||
AK146941.1 | - | 8 | 853 | |||
AK151628.1 | - | 8 | 819 | |||
AK191866.1 | - | 1 | 62 | |||
BC029077.1 | - | 7 | 669 | 4019292 | ||
BC059942.1 | - | 8 | 855 | 6529685 | ||
BC062178.1 | - | 8 | 812 | 6330944 | ||
U34960.1 | - | 7 | 649 | |||
U38505.1 | - | 1 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
