siRNA Details
Gene Symbol: Gng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038655.1 | NP_001033744.1 | 3 | 283 | ||
NM_010319.3 | NP_034449.2 | 3 | 220 | |||
GenBank mRNA | AK141541.1 | - | 3 | 192 | ||
AK147843.1 | - | 3 | 183 | |||
AK158963.1 | - | 3 | 283 | |||
AY035844.1 | - | 3 | 203 | |||
BC034108.1 | - | 2 | 147 | 4503666 | ||
BY271138.1 | - | 3 | 240 | |||
U38499.1 | - | 1 | 35 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
