siRNA Details
Gene Symbol: Gngt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038664.2 | NP_001033753.2 | 3, 4 | 314 | ||
NM_001284393.1 | NP_001271322.1 | 3, 4 | 318 | |||
NM_001284397.1 | NP_001271326.1 | 3, 4 | 315 | |||
NM_023121.2 | NP_075610.1 | 3, 4 | 299 | |||
GenBank mRNA | AA028742.1 | - | 3, 4 | 269 | ||
AA423571.1 | - | 3, 4 | 188 | |||
AI159118.1 | - | 3, 4 | 256 | |||
AK010554.1 | - | 2, 3 | 1328 | |||
BC061005.1 | - | 3, 4 | 264 | 30295552 | ||
BE847751.1 | - | 3, 4 | 304 | |||
BF470980.1 | - | 2, 3 | 128 | |||
BY117112.1 | - | 3, 4 | 299 | |||
U38500.1 | - | 1, 2 | 21 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
