siRNA Details
Gene Symbol: Gp1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010326.2 | NP_034456.2 | 2 | 1014 | ||
GenBank mRNA | BC137765.1 | - | 2 | 994 | 8860784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
