siRNA Details
Gene Symbol: Lrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081088.1 | NP_001074557.1 | 9 | 1121 | ||
GenBank mRNA | AK049346.2 | - | 9 | 1084 | ||
AK084165.1 | - | 9 | 1085 | |||
AK165434.1 | - | 9 | 1083 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
