siRNA Details
Gene Symbol: Pdpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290822.1 | NP_001277751.1 | 1, 2 | 385 | ||
NM_010329.3 | NP_034459.2 | 1, 2 | 385 | |||
GenBank mRNA | AJ250246.1 | - | 1, 2 | 114 | ||
AJ297944.2 | - | 1, 2 | 91 | |||
AK158855.1 | - | 1, 2 | 385 | |||
BC026551.1 | - | 1, 2 | 269 | 4985288 | ||
CX240531.1 | - | 1, 2 | 284 | |||
M73748.1 | - | 1, 2 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
