siRNA Details
Gene Symbol: Gp49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291892.1 | NP_001278821.1 | 3 | 364 | ||
NM_008147.2 | NP_032173.1 | 3 | 359 | |||
GenBank mRNA | AK155788.1 | - | 1 | 614 | ||
AK167751.1 | - | 3 | 280 | |||
AK170384.1 | - | 3 | 296 | |||
AK170836.1 | - | 2 | 502 | |||
AK171133.1 | - | 3 | 301 | |||
AK171229.1 | - | 3 | 301 | |||
AK171573.1 | - | 3 | 308 | |||
BY204344.1 | - | 3 | 235 | |||
M65027.1 | - | 3 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
